Learn More
Invitrogen™ Annexin A10 Monoclonal Antibody (K94033_12H11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA555862
Description
Sequence of this protein is as follows: SMYGRDLIGD MREQLSDHFK DVMAGLMYPP PLYDAHELWH AMKGVGTDEN CLIEILASRT NGEIFQMREA YCLQYSNNLQ EDIYSETSGH FRDTLMNLVQ GTREEGYTDP AMAAQDAMVL WEACQQKTGE HKTMLQMILC NKSYQQLRLV FQEFQNISGQ DMVDAINECY DGYFQELLVA IVLCVRDKPA YFAYRLYSAI HDFGFHNKTV IRILIARSEI DLLTIRKRYK ERYGKSLFHD IRNFASGHYK
This gene encodes a member of the annexin family. Members of this calcium-dependent phospholipid-binding protein family play a role in the regulation of cellular growth and in signal transduction pathways. The function of this gene has not yet been determined.
Specifications
| Annexin A10 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| Q9UJ72 | |
| ANXA10 | |
| Recombinant human ANXA10 (immunogen range is 61-310). | |
| 50 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| K94033_12H11 | |
| Unconjugated | |
| ANXA10 | |
| annexin 14; annexin A10; Annexin-10; annexin-14; ANX14; Anxa10; RGD1560956 | |
| Mouse | |
| Protein A | |
| RUO | |
| 11199 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.