missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 55018-1-AP-150UL
This item is not returnable.
View return policy
Description
ANXA1 is a pleiotrophic protein produced by many cell types including peripheral blood leucocytes. The antibody is specific to ANXA1.
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.Specifications
| Annexin A1 | |
| Polyclonal | |
| Unconjugated | |
| ANXA1 | |
| Annexin 1, annexin A1, Annexin I, ANX1, ANXA1, Calpactin 2, Calpactin II, Chromobindin 9, Lipocortin I, LPC1, p35 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 16952, 25380, 301 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 0.26 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P04083, P07150, P10107 | |
| Anxa1 | |
| Peptide | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction