missing translation for 'onlineSavingsMsg'
Learn More

Annexin A1 Rabbit anti-Human, Clone: RM424, Novus Biologicals™

Product Code. 18310082 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18310082 25 μg 25µL
18365223 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18310082 Supplier Novus Biologicals Supplier No. NBP30917725UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Annexin A1 Monoclonal specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone RM424
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:100-1:200
Formulation PBS, 50% Glycerol, 1% BSA
Gene Alias annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Host Species Rabbit
Immunogen A peptide corresponding to the N-terminus of human Annexin A1
Purification Method Protein A purified
Quantity 25 μg
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 301
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.