missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Annexin A1 Mouse anti-Human, Clone: CPTC-ANXA1-1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07714-100ug
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal specifically detects Annexin A1 in Human samples. It is validated for Western Blot, Immunoprecipitation, Immunohistochemistry-Paraffin.
Specifications
| Annexin A1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 to 2 μg/mL, Immunoprecipitation 2-4 ug/mg protein, Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein | |
| Recombinant full-length human Annexin A1 protein | |
| 100 μg | |
| Protein Kinase, Signal Transduction | |
| 301 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| CPTC-ANXA1-1 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction