Learn More
Invitrogen™ Annexin A1 Monoclonal Antibody (6B7F8)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549198
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Human Annexin A1 shares 88% and 89% amino acid (aa) sequence identity with mouse and rat Annexin A1, respectively. Positive Control - WB: human A549 whole cell, human K562 whole cell, human U-87 MG whole cell. IHC: human colorectal adenocarcinoma tissue, human lung squamous cell carcinoma tissue, human placenta tissue. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.
Specifications
| Annexin A1 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and no preservative | |
| P04083 | |
| ANXA1 | |
| E.coli-derived human Annexin A1 recombinant protein (Position: A2-N346). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 6B7F8 | |
| Unconjugated | |
| ANXA1 | |
| ANAX1; Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; ANX1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; Phospholipase A2 inhibitory protein; RP11-71A24.1 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 301 | |
| -20°C | |
| Lyophilized |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.