missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Annexin A1 Monoclonal Antibody (2#)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA543959
This item is not returnable.
View return policy
Description
Annexin A1 Monoclonal Antibody for Western Blot
Annexin I belongs to a family of Ca (2+) -dependent phospholipid binding proteins that are preferentially located on the cytosolic face of the plasma membrane. Annexin I has a molecular mass of 40 kDa, with phospholipase A2 inhibitory activity. Since phospholipase A2 is required for the biosynthesis of the potent mediators of inflammation, prostaglandins and leukotrienes, annexin I may have potential anti-inflammatory activity.
Specifications
| Annexin A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P04083, P07150, P10107 | |
| ANXA1 | |
| Recombinant protein Annexin A1. The antigen corresponds to amino acid range 1-346 of the target protein. | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 κ |
| Western Blot | |
| 2# | |
| Unconjugated | |
| ANXA1 | |
| ANAX1; Annexin 1 (p35) (Lipocortin 1); annexin A1; Annexin I; annexin I (lipocortin I); annexin-1; ANX1; Anx-1; Anxa1; Anx-A1; C430014K04Rik; Calpactin II; calpactin-2; Chromobindin-9; lipocortin 1; lipocortin I; LPC1; Lpc-1; p35; Phospholipase A2 inhibitory protein; RP11-71A24.1 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 16952, 25380, 301 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction