Learn More
Annexin A1 Antibody (CL0199) [Alexa Fluor™ 405], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-52875AF405
Product Code. 30296088
  • £558.00 / 0.10mL
Description
Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Annexin A1
Monoclonal
Alexa Fluor 405
annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
This antibody was developed using a recombinant protein derived from P04083, with the exact immunogen sequence remaining proprietary.
0.1 mL
Protein Kinase, Signal Transduction
301
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence
CL0199
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified