missing translation for 'onlineSavingsMsg'
Learn More

Annexin A1 Antibody (ANXA1/6452R), DyLight 680, Novus Biologicals™

Product Code. 18346716 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18346716 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18346716 Supplier Novus Biologicals Supplier No. NBP314056FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Annexin A1 Monoclonal antibody specifically detects Annexin A1 in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Annexin A1
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone ANXA1/6452R
Conjugate DyLight 680
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias annexin A1, Annexin I, annexin I (lipocortin I), Annexin-1, ANX1, Calpactin II, calpactin-2, Chromobindin-9, Lipocortin I, LPC1annexin-1, p35, Phospholipase A2 inhibitory protein
Host Species Rabbit
Immunogen Recombinant full-length human Annexin A1 protein (Uniprot: P04083)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Protein Kinase, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 301
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.