missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKZF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61588
This item is not returnable.
View return policy
Description
ANKZF1 Polyclonal specifically detects ANKZF1 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| ANKZF1 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| ankyrin repeat and zinc finger domain containing 1, ankyrin repeat and zinc finger domain-containing protein 1, FLJ13144, Zinc finger protein 744, ZNF744FLJ10415 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 55139 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500 | |
| NP_060559.2 | |
| ANKZF1 | |
| The immunogen recognized by this antibody maps to a region between residue 1 and 50 of human ankyrin repeat and zinc finger domain containing 1 using the numbering given in entry NP_060559.2 (GeneID 55139). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction