missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKZF1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61589B
This item is not returnable.
View return policy
Description
ANKZF1 Polyclonal antibody specifically detects ANKZF1 in Human samples. It is validated for Western Blot
Specifications
| ANKZF1 | |
| Polyclonal | |
| Western Blot | |
| ankyrin repeat and zinc finger domain containing 1, ankyrin repeat and zinc finger domain-containing protein 1, FLJ13144, Zinc finger protein 744, ZNF744FLJ10415 | |
| The immunogen recognized by this antibody maps to a region between residue 676 and 726 of human ankyrin repeat and zinc finger domain containing 1 using the numbering given in entry NP_060559.2 (GeneID 55139). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55139 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction