missing translation for 'onlineSavingsMsg'
Learn More

Ankyrin Repeat Domain 33B Antibody, R&D Systems™

Product Code. 18614674 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18614674 50 μg 50µL
18627742 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18614674 Supplier Novus Biologicals Supplier No. NBP29801350ul
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Rabbit Polyclonal Antibody

Ankyrin Repeat Domain 33B Polyclonal specifically detects Ankyrin Repeat Domain 33B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence/Immunohistochemistry/Immunohistochemistry (Paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ankyrin Repeat Domain 33B
Applications Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/ Immunofluorescence 1:100-1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH 7.0)
Gene Alias ANKRD33B, Ankyrin Repeat Domain-Containing Protein 33B, Ankyrin Repeat Domain-Containing Protein LOC651746
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human Ankyrin Repeat Domain 33B fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 651746
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Product Line Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.