Learn More
Invitrogen™ Ankyrin G Monoclonal Antibody (N106/20), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545540
Description
1 μg/mL of MA5-45540 was sufficient for detection of Ankyrin-G in 20 μg of rat brain membrane lysate and assayed by colorimetric immunoblot analysis using goat anti-mouse IgG:HRP as the secondary antibody. Detects >approximately 200kDa. This antibody was formerly sold as clone S106-20.
Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintenance and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.
Specifications
| Ankyrin G | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| G5E8K5, O70511, Q12955 | |
| ANK3 | |
| Fusion protein 1000 C-terminal amino acids of human Ankyrin G (also known as ANK-3 or ankyrin-3) encompassing all of Ankyrin G with the exception of Ankyrin repeats (aa 3377-4377). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| N106/20 | |
| APC | |
| ANK3 | |
| 2900054D09Rik; AI314020; ANK3; ANK-3; AnkG; ankyrin 3; ankyrin 3 (G); ankyrin 3, epithelial; ankyrin 3, node of Ranvier (ankyrin G); ankyrin G; ankyrin-3; Ankyrin-G; brain-specific ankyrin-G; LOW QUALITY PROTEIN: ankyrin-3; MRT37 | |
| Mouse | |
| Protein G | |
| RUO | |
| 11735, 288, 361833 | |
| 4°C | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.