missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin 3 Antibody (S106-20), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59310-25ug
This item is not returnable.
View return policy
Description
Ankyrin 3 Monoclonal specifically detects Ankyrin 3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Ankyrin 3 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 50% glycerol with 0.09% Sodium Azide | |
| ANK3 | |
| Fusion protein 1000 C-terminal amino acids of human Ankyrin G encompassing all of Ankyrin G with the exception of Ankyrin repeats | |
| 25 μg | |
| Primary | |
| Detects >200kDa. | |
| Store at -20°C. | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| S106-20 | |
| Western Blot 1:100 - 1:2000, Immunocytochemistry/Immunofluorescence 1:10 - 1:500 | |
| ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 288 | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction