missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin 3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP3-05549MFV450
This item is not returnable.
View return policy
Description
Ankyrin 3 Polyclonal antibody specifically detects Ankyrin 3 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Ankyrin 3 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier | |
| The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli. | |
| 0.1 mL | |
| Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers | |
| 288 | |
| Store at 4C in the dark. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Chicken | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction