missing translation for 'onlineSavingsMsg'
Learn More

Ankyrin 3 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18407488 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407488 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407488 Supplier Novus Biologicals Supplier No. NBP305549AF488

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chicken Polyclonal Antibody

Ankyrin 3 Polyclonal antibody specifically detects Ankyrin 3 in Human, Mouse, Rat, Bovine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ankyrin 3
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias ankyrin G, Ankyrin G119, ankyrin-3, ankyrin-G, Node Of Ranvier
Host Species Chicken
Immunogen The C-terminal 398 amino acids of human Ankyrin 3 expressed in and purified from E. coli.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Signaling, Neuronal Cell Markers, Neuroscience, Plasma Membrane Markers
Primary or Secondary Primary
Gene ID (Entrez) 288
Target Species Human, Mouse, Rat, Bovine, Monkey
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgY
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.