missing translation for 'onlineSavingsMsg'
Learn More

Ankyrin 1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA542203

Code : NEW

Additional Details : Weight : 0.00010kg

Product Code. 15595312

  • £403.00 / 100µL
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Peptide sequence: VVRQIDLSSA DAAQEHEEVE LRGSGLQPDL IEGRKGAQIV KRASLKRGKQ Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%

Ankyrin 1 Polyclonal antibody specifically detects Ankyrin 1 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifications

Specifications

Ankyrin 1
Polyclonal
Unconjugated
ANK1
ANK; ANK1; ANK-1; ANK-1; erythrocyte skeletal protein; ankyrin 1; ankyrin 1, erythrocytic; ankyrin 1, erythroid; ankyrin-1; Ankyrin-R; erythrocyte ankyrin; nb; normoblastic anemia; pale; rCG_43073; SPH1; SPH2
Rabbit
Protein A
RUO
286
-20° C, Avoid Freeze/Thaw Cycles
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 2% sucrose and 0.09% sodium azide
P16157
ANK1
Synthetic peptide directed towards the C-terminal of human ANK1
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers