missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ankyrin 1 Antibody (S388A-60), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59316AF488
This item is not returnable.
View return policy
Description
Ankyrin 1 Monoclonal specifically detects Ankyrin 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| Ankyrin 1 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ANK1 | |
| Fusion protein amino acids 1-1881 (full-length) of human Ankyrin-R | |
| 100 μL | |
| 286 | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| S388A-60 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ANK, ANK-1, ankyrin 1, erythrocytic, ankyrin-1, Ankyrin-R, Erythrocyte ankyrin, SPH1, SPH2 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| Detects 200kDa. Cross-reacts with Ankyrin-B and Ankyrin-G. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction