missing translation for 'onlineSavingsMsg'
Learn More

Ankyrin 1 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18406862 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406862 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406862 Supplier Novus Biologicals Supplier No. NBP171805FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Ankyrin 1 Polyclonal antibody specifically detects Ankyrin 1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Ankyrin 1
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 680
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ANK, ANK-1, ankyrin 1, erythrocytic, ankyrin-1, Ankyrin-R, Erythrocyte ankyrin, SPH1, SPH2
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1831 and 1881 of human Ankyrin 1 using the numbering given in entry NP_065209.2 (GeneID 286).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.