missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
ANKS6 Polyclonal specifically detects ANKS6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
Specifications
| Antigen | ANKS6 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Janelia Fluor 549 |
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin |
| Formulation | 50mM Sodium Borate |
| Gene Alias | ANKRD14, ankyrin repeat and SAM domain-containing protein 6, ankyrin repeat and sterile alpha motif domain containing 6, ankyrin repeat domain 14, Ankyrin repeat domain-containing protein 14, DKFZp686D24121, DKFZp781I0117, FLJ36928, MGC70366, SAM domain-containing protein 6, SamCystin, SAMD6, sterile alpha motif domain containing 6, Sterile alpha motif domain-containing protein 6 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human ANKS6 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?