missing translation for 'onlineSavingsMsg'
Learn More

ANKS6 Antibody, Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18349235 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18349235 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18349235 Supplier Novus Biologicals Supplier No. NBP306064AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKS6 Polyclonal specifically detects ANKS6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKS6
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias ANKRD14, ankyrin repeat and SAM domain-containing protein 6, ankyrin repeat and sterile alpha motif domain containing 6, ankyrin repeat domain 14, Ankyrin repeat domain-containing protein 14, DKFZp686D24121, DKFZp781I0117, FLJ36928, MGC70366, SAM domain-containing protein 6, SamCystin, SAMD6, sterile alpha motif domain containing 6, Sterile alpha motif domain-containing protein 6
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human ANKS6 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Autophagy, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Transcription Factors and Regulators, Vision, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 203286
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.