missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKS1B Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£157.00 - £358.00
Specifications
| Antigen | ANKS1B |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
ANKS1B Polyclonal antibody specifically detects ANKS1B in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| ANKS1B | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Alzheimers Research, Neurodegeneration, Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 56899 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| AIDA-1AIDA, amyloid-beta precursor protein intracellular domain associated protein 1, Amyloid-beta protein intracellular domain-associated protein 1, ANKS2, ankyrin repeat and sterile alpha motif domain containing 1B, ankyrin repeat and sterile alpha motif domain-containing protein 1B, cajalin 2, cajalin-2, E2A-PBX1-associated protein, EB1, EB-1MGC26087 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 261-510 of human ANKS1B (NP_858056.2). GFPFSAIHQVHNTGDWGEPSITLRPPNEATASTPVQYWQHHPEKLIFQSCDYKAFYLGSMLIKELRGTESTQDACAKMRANCQKSTEQMKKVPTIILSVSYKGVKFIDATNKNIIAEHEIRNISCAAQDPEDLSTFAYITKDLKSNHHYCHVFTAFDVNLAYEIILTLGQAFEVAYQLALQARKGGHSSTLPESFENKPSKPIPKPRVSIRKSVQIDPSEQKTLANLPWIVEPGQEAKRGINTKYETTIF | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title