missing translation for 'onlineSavingsMsg'
Learn More

ANKS1A Antibody, Alexa Fluor™ 405, Novus Biologicals™

Product Code. 18414434 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414434 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414434 Supplier Novus Biologicals Supplier No. NBP171806AF405

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKS1A Polyclonal antibody specifically detects ANKS1A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKS1A
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ANKS1ankyrin repeat and SAM domain-containing protein 1A, ankyrin repeat and SAM domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1, ankyrin repeat and sterile alpha motif domain containing 1A, KIAA0229MGC42354, ODIN
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 875 and 925 of human Ankyrin Repeat and Sterile Apha Motif Domain Containing 1A using the numbering given in entry NP_056060.2 (GeneID 23294).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23294
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.