missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569882
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 92%; Dog: 92%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rat: 100%.
ANKRD9 gene ontology annotations related to this gene include post-translational protein modification; protein localization to plasma membrane.
Specifications
| ANKRD9 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD9 | |
| 2500003O20Rik; Ankrd9; ankyrin repeat domain 9; ankyrin repeat domain-containing protein 9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 122416 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96BM1 | |
| ANKRD9 | |
| synthetic peptide directed towards the middle region of human ANKRD9. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction