missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ANKRD7 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA583660

Product Code. 16350615

  • £478.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RTALILAVSG EPPCLVKLLL QQGVELCYEG IVDSQLRNMF ISMVLLHRYP QFTASHGKKK HA Highest antigen sequence indentity to the following orthologs: Mouse - 34%, Rat - 37%.

ANKRD7 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANKRD7
Polyclonal
Unconjugated
ANKRD7
ANKRD7; ankyrin repeat domain 7; ankyrin repeat domain-containing protein 7; testicular tissue protein Li 19; testis-specific ankyrin motif containing protein; testis-specific protein TSA806; TSA806
Rabbit
Antigen affinity chromatography
RUO
56311
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q92527
ANKRD7
Recombinant protein corresponding to Human ANKRD7. Recombinant protein control fragment (Product #RP-100368).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.