missing translation for 'onlineSavingsMsg'
Learn More

ANKRD7 Antibody, Novus Biologicals™

Product Code. 18354359 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18354359 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18354359 Supplier Novus Biologicals Supplier No. H00056311B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ANKRD7 Polyclonal antibody specifically detects ANKRD7 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD7
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. AAH32799.1
Gene Alias ankyrin repeat domain 7, ankyrin repeat domain-containing protein 7, testis-specific ankyrin motif containing protein, Testis-specific protein TSA806, TSA806
Host Species Mouse
Immunogen ANKRD7 (AAH32799.1, 1 a.a. - 201 a.a.) full-length human protein. MQDKKYRTPLHLACANGHTDVVLFLIEQQCKINVRDSENKSPLIKAVQCQNEDCATILLNFGADPDLRDIRYNTVLHYAVCGQSLSLVEKLLEYEADLEAKNKDGYTPLLVAVINNNPKMVKFLLEKGADVNASDNYQRTALILAVSGEPPCLVKLLLQQGVELCYEGIVDSQLRNMFISMVLLHRYPQFTASHGKKKHAK
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56311
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.