missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | ANKRD7 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ANKRD7 Polyclonal specifically detects ANKRD7 in Human samples. It is validated for Western Blot.Specifications
| ANKRD7 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| ankyrin repeat domain 7, ankyrin repeat domain-containing protein 7, testis-specific ankyrin motif containing protein, Testis-specific protein TSA806, TSA806 | |
| ANKRD7 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Q92527 | |
| 56311 | |
| Synthetic peptides corresponding to ANKRD7(ankyrin repeat domain 7) The peptide sequence was selected from the middle region of ANKRD7. Peptide sequence SENKSPLIKAVQCQNEDCATILLNFGADPDLRDIRYNTVLHYAVCGQSLS. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title