missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD53 Antibody (OTI1E1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-72217CL1
This item is not returnable.
View return policy
Description
ANKRD53 Monoclonal antibody specifically detects ANKRD53 in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF,Immunocytochemistry/Immunofluorescence
Specifications
| ANKRD53 | |
| Monoclonal | |
| CoraFluor 1 | |
| ankyrin repeat domain 53, ankyrin repeat domain-containing protein 53, FLJ12056, FLJ36160 | |
| Human recombinant protein fragment corresponding to amino acids 1-300 of human ANKRD53 (NP_079209) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF, Immunocytochemistry/Immunofluorescence | |
| OTI1E1 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 79998 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction