missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD52 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92833-0.02ml
This item is not returnable.
View return policy
Description
ANKRD52 Polyclonal antibody specifically detects ANKRD52 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ANKRD52 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| ANKRD33, ankyrin repeat domain 33, ankyrin repeat domain 52, ankyrin repeat domain-containing protein 52, CVWG5837, FLJ34236, PP6-ARS-C, protein phosphatase 6 ankyrin repeat subunit C, serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit C | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 450-660 of human ANKRD52 (NP_775866.2). RRRDKFGRTPLHYAAANGSYQCAVTLVTAGAGVNEADCKGCSPLHYAAASDTYRRAEPHTPSSHDAEEDEPLKESRRKEAFFCLEFLLDNGADPSLRDRQGYTAVHYAAAYGNRQNLELLLEMSFNCLEDVESTIPVSPLHLAAYNGHCEALKTLAETLVNLDVRDHKGRTALFLATERGSTECVEVLTAHGASALIKERKRKWTPLHAAA | |
| 0.02 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 283373 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction