missing translation for 'onlineSavingsMsg'
Learn More

ANKRD50 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18400724 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400724 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400724 Supplier Novus Biologicals Supplier No. NBP150032AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKRD50 Polyclonal antibody specifically detects ANKRD50 in Human, Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD50
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ankyrin repeat domain 50
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1379 and 1429 of human Ankyrin Repeat Domain 50 using the numbering given in entry NP_065070.1 (GeneID 57182).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57182
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.