Learn More
ANKRD45 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA556809
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: SQQEEENEEE EAQEPEETGP KNPLLQPALT GDVEGLQKIF EDPENPHHEQ AMQLLLEEDI VGRNLLYAAC MAGQSDVIRA LAKYGVNLN Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 29%.
ANKRD45 Polyclonal antibody specifically detects ANKRD45 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
ANKRD45 | |
Polyclonal | |
Unconjugated | |
ANKRD45 | |
4933409K03Rik; ANKRD45; ankyrin repeat domain 45; ankyrin repeat domain-containing protein 45; cancer/testis antigen 117; CT117; RGD1311153; RP3-436N22.4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
339416 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TZF3 | |
ANKRD45 | |
Recombinant protein corresponding to Human ANKRD45 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only