missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD42 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569122
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 93%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.
ANKRD42 is a protein coding gene.
Specifications
| ANKRD42 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD42 | |
| 4931426M20; 4933417L02Rik; Ankrd42; ankyrin repeat domain 42; ankyrin repeat domain-containing protein 42; Ikbn; PPP1R79; protein phosphatase 1, regulatory subunit 79; RGD1310789; SARP; several ankyrin repeat protein | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 338699 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8N9B4 | |
| ANKRD42 | |
| synthetic peptide directed towards the N terminal region of human ANKRD42. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction