Learn More
Invitrogen™ ANKRD28 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109392
Description
Immunogen sequence: MAFLKLRDQP SLVQAIFNGD PDEVRALIFK KEDVNFQDNE KRTPLHAAAY LGDAEIIELL ILSGARVNAK DSKWLTPLHR AVASCSEEAV QVLLKHSADV NARDKNWQTP LHIAAANKAV KCAEALVPLL SNVNVSDRAG RTALHHAAFS GHGEMVKLLL SRGANINAFD KKDRRAIHWA AYMGHIEVVK LLVSHGAEVT CKDKKSYTPL HAAASSGMIS VVKYLLDLGV DMNEPNAYGN TPLHVACYNG QDVVVNELID CGAIVNQKNE KGFTPLHFAA ASTHGALCLE LLVGNGADVN.
Putative regulatory subunit of protein phosphatase 6 (PP6) that may be involved in the recognition of phosphoprotein substrates. Involved in the PP6-mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha. Selectively inhibits the phosphatase activity of PPP1C. Targets PPP1C to modulate HNRPK phosphorylation. ANKRD28 selectively regulated phosphorylation of hnRNP-K at ser284, an effect that was enhanced in vivo by an S1013A/S1017A double mutant, suggesting that ANKRD28 targets PP1 in a regulated manner to dephosphorylate hnRNP-K selectively at ser284.
Specifications
| ANKRD28 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD28 | |
| AI465466; AI604979; ANKRD28; ankyrin repeat domain 28; ankyrin repeat domain-containing protein 28; E430019N21Rik; KIAA0379; mKIAA0379; phosphatase interactor targeting K protein; phosphatase interactor targeting protein hnRNP K; PITK; PP6-ARS-A; PPP1R65; protein phosphatase 1, regulatory subunit 65; protein phosphatase 6 ankyrin repeat subunit A; RGD1559931; serine/threonine-protein phosphatase 6 regulatory ankyrin repeat subunit A; serine/threonine-protein phosphatase 6 regulatory subunit ARS-A | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 105522, 23243 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.58 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O15084, Q505D1 | |
| ANKRD28 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ANKRD28 (NP_056014.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.