missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114462
This item is not returnable.
View return policy
Description
Positive controls that may be used include: K562 Cell Lysate.
May be a guanine exchange factor (GEF) for Rab21, Rab32 and Rab38 and regulate endosome dynamics (PubMed:16525121, PubMed:18477474). May regulate the participation of VAMP7 in membrane fusion events; in vitro inhibits VAMP7-mediated SNARE complex formation by trapping VAMP7 in a closed, fusogenically inactive conformation (PubMed:23104059). Involved in peripheral melanosomal distribution of TYRP1 in melanocytes; the function, which probably is implicating vesicle-trafficking, includes cooperation with Rab32, Rab38 and VAMP7. Involved in the regulation of neurite growth; the function seems to require its GEF activity, probably towards Rab21, and VAMP7 but not Rab32/38. Proposed to be involved in Golgi sorting of VAMP7 and transport of VAMP7 vesicles to the cell surface; the function seems to implicate kinesin heavy chain isoform 5 proteins, GOLGA4, RAB21 and MACF1 (PubMed:22705394). Required for the colocalization of VAMP7 and Rab21, probably on TGN sites (PubMed:19745841). Involved in GLUT1 endosome-to-plasma membrane trafficking; the function is dependent of association with VPS29 (PubMed:24856514). Regulates the proper trafficking of melanogenic enzymes TYR, TYRP1 and DCT/TYRP2 to melanosomes in melanocytes. [UniProt].
Specifications
| ANKRD27 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD27 | |
| ANKRD27; ankyrin repeat domain 27; ankyrin repeat domain 27 (VPS9 domain); ankyrin repeat domain-containing protein 27; PP12899; VARP; Vps9 domain and ankyrin-repeat-containing protein; VPS9 domain-containing protein; VPS9-ankyrin-repeat protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 84079 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q96NW4 | |
| ANKRD27 | |
| VARP antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human VARP. The immunogen is located within amino acids 50 - 100 of VARP. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction