missing translation for 'onlineSavingsMsg'
Learn More

ANKRD27 Antibody, PerCP, Novus Biologicals™

Product Code. 30032343 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30032343 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30032343 Supplier Novus Biologicals Supplier No. NBP150030PCP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKRD27 Polyclonal antibody specifically detects ANKRD27 in Human samples. It is validated for Western Blot, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD27
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias ankyrin repeat domain 27 (VPS9 domain), ankyrin repeat domain-containing protein 27, DKFZp434L0718, FLJ00040, VARP, Vps9 domain and ankyrin-repeat-containing protein, VPS9 domain-containing protein, VPS9-ankyrin-repeat protein
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 1000 and 1050 of human Vps9 Domain and Ankyrin-Repeat-Containing Protein using the numbering given in entry NP_115515.2 (GeneID 84079).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 84079
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.