missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD27 Antibody, Alexa Fluor™ 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82072AF350
This item is not returnable.
View return policy
Description
ANKRD27 Polyclonal antibody specifically detects ANKRD27 in Human samples. It is validated for Western Blot, ELISA
Specifications
| ANKRD27 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| Alexa Fluor 350 | |
| ankyrin repeat domain 27 (VPS9 domain), ankyrin repeat domain-containing protein 27, DKFZp434L0718, FLJ00040, VARP, Vps9 domain and ankyrin-repeat-containing protein, VPS9 domain-containing protein, VPS9-ankyrin-repeat protein | |
| ANKRD27 antibody was raised against a 16 amino acid synthetic peptide near the amino terminus of human ANKRD27. The immunogen is located within amino acids 50 - 100 of ANKRD27. Amino Acid Squence: HFQTLNGKDVFIQGNR | |
| 0.1 mL | |
| Signal Transduction | |
| 84079 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu