missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114852
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ANKRD26.
ANKRD26 encodes a protein containing N-terminal ankyrin repeats which function in protein-protein interactions. Mutations in this gene are associated with autosomal dominant thrombocytopenia-2. Pseudogenes of this gene are found on chromosome 7, 10, 13 and 16. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
| ANKRD26 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD26 | |
| 5730521P14Rik; ANKRD26; ankyrin repeat domain 26; ankyrin repeat domain-containing protein 26; bA145E8.1; BC008111; KIAA1074; mKIAA1074; RGD1310736; THC2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22852, 232339, 312667 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q811D2, Q9UPS8 | |
| ANKRD26 | |
| A synthesized peptide derived from human ANKRD26(Accession Q9UPS8), corresponding to amino acid residues S793-L843. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction