Learn More
ANKRD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559339
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IIKLLLLHGA DMMTKNLAGK TPTDLVQLWQ ADTRHALEHP EPGAEHNGLE GPNDSGRETP QPVPAQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
ANKRD2 may play an important role in skeletal muscle hypertrophy.Specifications
ANKRD2 | |
Polyclonal | |
Unconjugated | |
Ankrd2 | |
Ankrd2; ankyrin repeat domain 2; ankyrin repeat domain 2 (stretch responsive muscle); ankyrin repeat domain-containing protein 2; ankyrin repeat domain-containing protein 2; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 2; ARPP; hArpp; LOW QUALITY PROTEIN: ankyrin repeat domain-containing protein 2; mArpp; Skeletal muscle ankyrin repeat protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
26287 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry, Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9GZV1 | |
Ankrd2 | |
Recombinant protein corresponding to Human ANKRD2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only