missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117433
This item is not returnable.
View return policy
Description
Centrifuge briefly prior to opening.
Required to prevent the misactivation of serine (Ser) with tRNA(Ala) by promoting the hydrolysis of Ser-mischarged tRNA(Ala), thereby playing a role in translational fidelity. Binds directly to the catalytic domain of AARS/AlaRS and captures Ser that is misactivated by AARS/AlaRS, preventing the charging of Ser adenylates to tRNA(Ala) and precluding Ser misincorporation in nascent peptides. [UniProt].
Specifications
| ANKRD16 | |
| Polyclonal | |
| Unconjugated | |
| ANKRD16 | |
| 2810455F06Rik; AI646698; Ankrd16; ankyrin repeat domain 16; ankyrin repeat domain-containing protein 16; D430029B21Rik; hypothetical protein LOC550225; si:dkey-108d22.7; Unknown (protein for MGC:143369); zgc:110558 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 320816 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.63 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| A2AS55 | |
| ANKRD16 | |
| Recombinant protein encompassing a sequence within the N-terminus region of mouse ANKRD16. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction