missing translation for 'onlineSavingsMsg'
Learn More

ANKRD15 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18420658 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18420658 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18420658 Supplier Novus Biologicals Supplier No. NBP103345C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ANKRD15 Polyclonal antibody specifically detects ANKRD15 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKRD15
Applications Western Blot
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias ANKRD15ankyrin repeat domain 15, Ankyrin repeat domain-containing protein 15, KANKDKFZp451G231, KIAA0172MGC43128, Kidney ankyrin repeat-containing protein, KN motif and ankyrin repeat domain-containing protein 1, KN motif and ankyrin repeat domains 1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 1302 and 1352 of human KN motif and ankyrin repeat domains 1 using the numbering given in entry NP_055973.2 (GeneID 23189).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 23189
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.