missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKRD13C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569537
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
Acts as a molecular chaperone for G protein-coupled receptors, regulating their biogenesis and exit from the ER.
Specifications
| ANKRD13C | |
| Polyclonal | |
| Unconjugated | |
| ANKRD13C | |
| AI505652; Ankrd13c; ankyrin repeat domain 13c; ankyrin repeat domain-containing protein 13C; AU022220; dJ677H15.3; fc57c01; RP4-677H15.5; similar to hypothetical protein DKFZp566D1346; wu:fc57c01; wu:fq56c05; zgc:56077 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 433667 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q3UX43 | |
| ANKRD13C | |
| synthetic peptide directed towards the C-terminal region of mouse ANKRD13C. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction