missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKRD1 Antibody, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-33368-100ul
This item is not returnable.
View return policy
Description
ANKRD1 Monoclonal antibody specifically detects ANKRD1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| ANKRD1 | |
| Monoclonal | |
| Western Blot 1:500 - 1:1000, ELISA Recommended starting concentration is 1 μg/mL | |
| ALRP, ankyrin repeat domain 1 (cardiac muscle), ankyrin repeat domain-containing protein 1, bA320F15.2, C193, C-193, Cardiac ankyrin repeat protein, CARPCytokine-inducible nuclear protein, CVARP, Cytokine-inducible gene C-193 protein, HA1A2, liver ankyrin repeat domain 1, MCARP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human ANKRD1 (NP_055206.2).,, Sequence:, MMVLKVEELVTGKKNGNGEAGEFLPEDFRDGEYEAAVTLEKQEDLKTLLAHPVTLGEQQWKSEKQREAELKKKKLEQRSKLENLEDLEIIIQLKKRKKYRKTKVPVVKEPEPEIITEPVDVPTFLKAALE | |
| 100 μL | |
| Cardiovascular Biology | |
| 27063 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction