missing translation for 'onlineSavingsMsg'
Learn More

ANKMY1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-97343JF549

Product Code. 18029676

  • £322.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ANKMY1 Polyclonal specifically detects ANKMY1 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ANKMY1
Polyclonal
Immunocytochemistry/Immunofluorescence
ankyrin repeat and MYND domain containing 1, DKFZp686D20155, TSAL1, ZMYND13
Produced in rabbits immunized with E. coli-derived Human ANKMY1 fragment.
0.1 mL
Primary
Human
Purified
Immunofluorescence
Janelia Fluor 549
50mM Sodium Borate
Rabbit
Antigen Affinity-purified
RUO
51281
Store at 4°C in the dark.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.