missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKLE2 Antibody - BSA Free, Novus Biologicals™
Chicken Polyclonal Antibody
£134.00 - £398.00
Specifications
| Antigen | ANKLE2 |
|---|---|
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18652626
|
Novus Biologicals
NBP2-41145-0.025mg |
0.025 mg |
£134.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18160199
|
Novus Biologicals
NBP2-41145 |
0.1 mg |
£398.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ANKLE2 Polyclonal specifically detects ANKLE2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA.Specifications
| ANKLE2 | |
| Western Blot, ELISA | |
| Unconjugated | |
| RUO | |
| PBS with 0.02% Sodium Azide | |
| 23141 | |
| Antibody was raised against a 19 amino acid synthetic peptide near the center of human ANKLE2. The immunogen is located within amino acids 280 - 330 of ANKLE2. Amino Acid Squence: CSESETVNKERANSYKNPRT | |
| Primary | |
| Three isoforms of ANKLE2 are known to exist; this antibody will detect all three. ANKLE2 antibody is predicted to not cross-react with ANKLE1. |
| 1 mg/mL | |
| Polyclonal | |
| Chicken | |
| Human, Mouse, Rat | |
| ankyrin repeat and LEM domain containing 2, ankyrin repeat and LEM domain-containing protein 2, FLJ22280, FLJ36132, KIAA0692, LEM domain containing 7, LEMD7 | |
| ANKLE2 | |
| IgY | |
| Affinity Purified | |
| 121 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title