missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKLE1 Polyclonal Antibody

Chicken Polyclonal Antibody
Brand: Invitrogen™ PA572735
This item is not returnable.
View return policy
Description
ANKLE1 Polyclonal Antibody for Western Blot, ELISA
Endonuclease that probably plays a role in the DNA damage response and DNA repair.
Specifications
| ANKLE1 | |
| Polyclonal | |
| Unconjugated | |
| ANKLE1 | |
| 8430438L13Rik; Ankle1; Ankrd41; ankyrin repeat and LEM domain containing 1; ankyrin repeat and LEM domain-containing protein 1; ankyrin repeat domain 41; ankyrin repeat domain-containing protein 41; C430003I23; LEM domain containing 6; Lem3; LEMD6; LEM-domain containing 3; LEM-domain containing protein 3; RGD1308184 | |
| Chicken | |
| Antigen affinity chromatography | |
| RUO | |
| 126549, 234396, 361122 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| A8VU90, Q8NAG6 | |
| ANKLE1 | |
| ANKLE1 antibody for a 15 amino acid synthetic peptide near the carboxy terminus of human ANKLE1. The immunogen is within the last 50 amino acids of ANKLE1. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgY |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction