missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANKK1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92306-0.1ml
This item is not returnable.
View return policy
Description
ANKK1 Polyclonal antibody specifically detects ANKK1 in Human samples. It is validated for Western Blot
Specifications
| ANKK1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| ankyrin repeat and kinase domain containing 1, ankyrin repeat and protein kinase domain-containing protein 1, EC 2.7.11, EC 2.7.11.1, PKK2, Protein kinase PKK2, SGK288, Sugen kinase 288, X-kinasesgK288 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human ANKK1 (NP_848605.1). MAADPTELRLGSLPVFTRDDFEGDWRLVASGGFSQVFQARHRRWRTEYAIKCAPCLPPDAASSDVNYLIEEAAKMKKIKFQHIVSIYGVCKQPLGIVMEFMANGSLEKVLSTHSLCWKLRFRIIHETSLAMNFLHSIKPPLLHLDLKPGNILLDSNMHVKISDFGLSKWMEQSTRMQYIERSALRGMLSYIPPEMFLESNKAPGPKYDVYSFAIVIWELLTQKKPYSGFNMMMIIIRVAAGMRPSLQPVSDQWPSEAQQMVDLMKRCWDQDPKKRPCFLDITIETDILLSLLQSRVAVPE | |
| 0.1 mL | |
| Signal Transduction | |
| 255239 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction