missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKHD1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA552595
This item is not returnable.
View return policy
Description
Immunogen sequence: QKVERQLQMK TQQQFTKEYL ETKGQKDTVS LHQQCSHRGV FPEGEGDGSL PEDHFSELPQ VDTILFKDND VDDEQQSPPS AEQIDFVPVQ PLSSPQCNFS SDLGSNGTNS LELQKVSGNQ QIVGQPQIAI TGHDQGLLVQ EPD Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 83%.
This gene encodes a protein with multiple ankyrin repeat domains and a single KH-domain. Co-transcription of this gene and the neighboring downstream gene (EIF4EBP3) generates a transcript (MASK-BP3) which encodes a fusion protein comprised of the MASK protein sequence for the majority of the protein and a different C-terminus due to an alternate reading frame for the EIF4EBP3 segments.
Specifications
| ANKHD1 | |
| Polyclonal | |
| Unconjugated | |
| ANKHD1 | |
| 1110004O12Rik; 4933432B13Rik; 9130019P20Rik; A530027J04Rik; A630021B20Rik; AA571404; ANKHD1; ankyrin repeat and KH domain containing 1; ankyrin repeat and KH domain-containing protein 1; HIV-1 Vpr-binding ankyrin repeat protein; hMASK; KIAA1085; MASK; MASK1; mFLJ00246; mKIAA1085; Multiple ankyrin repeats single KH domain; multiple ankyrin repeats, single KH-domain homolog; PP2500; VBARP | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54882 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IWZ3 | |
| ANKHD1 | |
| Recombinant protein corresponding to Human ANKHD1. Recombinant protein control fragment (Product #RP-102515). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction