missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKFY1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555096
This item is not returnable.
View return policy
Description
Immunogen sequence: YVFSQLLQCK QLDLELKDHE GSTALWLAVQ HITVSSDQSV NPFEDVPVVN GTSFDENSFA ARLIQRGSHT DAPDTATGNC LLQRAAGAGN EAAALFLATN Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 94%.
ANKFY1 encodes a cytoplasmic protein that contains a coiled-coil structure and a BTB/POZ domain at its N-terminus, ankyrin repeats in the middle portion, and a FYVE-finger motif at its C-terminus. This protein belongs to a subgroup of double zinc finger proteins which may be involved in vesicle or protein transport.
Specifications
| ANKFY1 | |
| Polyclonal | |
| Unconjugated | |
| ANKFY1 | |
| Ankfy1; ANKHZN; ankyrin repeat and FYVE domain containing 1; ankyrin repeat and FYVE domain-containing protein 1; ankyrin repeat hooked to a zinc finger motif; ankyrin repeat hooked to zinc finger motif; ankyrin repeats hooked to a zinc finger motif; BTBD23; KIAA1255; mKIAA1255; rabankyrin-5; Rank-5; ZFYVE14 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51479 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P2R3 | |
| ANKFY1 | |
| Recombinant protein corresponding to Human ANKFY1. Recombinant protein control fragment (Product #RP-93512). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction