missing translation for 'onlineSavingsMsg'
Learn More

ANKFY1 Antibody, Novus Biologicals™

Product Code. 18374639 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.05 mg
Unit Size:
0.05mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18374639 0.05 mg 0.05mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18374639 Supplier Novus Biologicals Supplier No. H00051479B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

ANKFY1 Polyclonal antibody specifically detects ANKFY1 in Human, Bovine samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen ANKFY1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence 10 μg/mL
Formulation PBS (pH 7.4)
Gene Accession No. AAI52992.1
Gene Alias ANKHZN, ankyrin repeat and FYVE domain containing 1, ankyrin repeat and FYVE domain-containing protein 1, ankyrin repeat hooked to zinc finger motif, ankyrin repeats hooked to a zinc finger motif, DKFZp686M19106, KIAA1255, Rabankyrin-5, ZFYVE14
Host Species Mouse
Immunogen ANKFY1 (AAI52992.1, 1 a.a. - 1170 a.a.) full-length human protein. MAEEEVAKLEKHLMLLRQEYVKLQKKLAETEKRCALLAAQANKESSSESFISRLLAIVADLYEQEQYSDLKIKVGDRHISAHKFVLAARSDSWSLANLSSTKELDLSDANPEVTMTMLRWIYTDELEFREDDVFLTELMKLANRFQLQLLRERCEKGVMSLVNVRNCIRFYQTAEELNASTLMNYCAEIIASHWDDLRKEDFSSMSAQLLYKMIKSKTEYPLHKAIKVEREDVVFLYLIEMDSQLPGKLNEADHNGDLALDLALSRRLESIATTLVSHKADVDMVDKSGWSLLHKGIQRGDLFAATFLIKNGAFVNAATLGAQETPLHLVALYSSKKHSADVMSEMAQIAEALLQAGANPNMQDSKGRTPLHVSIMAGNEYVFSQLLQCKQLDLELKDHEGSTALWLAVQHITVSSDQSVNPFEDVPVVNGTSFDENSFAARLIQRGSHTDAPDTATGNCLLQRAAGAGNEAAALFLATNGAHVNHRNKWGETPLHTACRHGLANLTAELLQQGANPNLQTEEALPLPKEAASLTSLADSVHLQTPLHMAIAYNHPDVVSVILEQKANALHATNNLQIIPDFSLKDSRDQTVLGLALWTGMHTIAAQLLGSGAAINDTMSDGQTLLHMAIQRQDSKSALFLLEHQADINVSRTQDGETALQLAIRNQLPLVVDAICTRGADMSVPDEKGNPPLWLALANNLEDIASTLVRHGCDATCWGPGPGGCLQTLLHRAIDENNEPTACFLIRSGCDVNSPRQPGANGEGEEEARDGQTPLHLAASWGLEETVQCLLEFGANVNAQDAEGRTPIHVAISSQHGVIIQLLVSHPDIHLNVRDRQGLTPFACAMTFKNNKSAEAILKRESGAAEQVDNKGRNFLHVAVQNSDIESVLFLISVHANVNSRVQDASKLTPLHLAVQAGSEIIVRNLLLAGAKVNELTKHRQTALHLAAQQDLPTICSVLLENGVDFAAVDENGNNALHLAVMHGRLNNIRVLLTECTVDAEAFNLRGQSPLHILGQYGKENAAAIFDLFLECMPGYPLDKPDADGSTVLLLAYMKGNANLCRAIVRSGARLGVNNNQGVNIFNYQVATKQLLFRLLDMLSKEPPWCDGSYCYECTARFGVTTRKHHCRHCGRLLCHKCSTKEIPIIKFDLNKPVRVCNICFDVLTLGGVS
Purification Method Protein A purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51479
Target Species Human, Bovine
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.