missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ANKEF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559453
This item is not returnable.
View return policy
Description
Immunogen sequence: DCCKYIAQRG CDLKWKNLDH KTPRAVAKEG GFKAASKEIR RAERIANKLA RPGAKNPNPL WALRLHDWSV EREAFLREAF AVLDRGDGSI SKNDFVMVLE Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
Specifications
| ANKEF1 | |
| Polyclonal | |
| Unconjugated | |
| ANKEF1 | |
| Ankef1; Ankrd5; ankyrin repeat and EF-hand domain containing 1; ankyrin repeat and EF-hand domain-containing protein 1; ankyrin repeat domain 5; ankyrin repeat domain-containing protein 5; AV276460 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 63926 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.6 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NU02 | |
| ANKEF1 | |
| Recombinant protein corresponding to Human ANKEF1. Recombinant protein control fragment (Product #RP-99427). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction