missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ANK3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27766-1-AP-150UL
This item is not returnable.
View return policy
Description
Ankyrin-G is found in several tissues including brain where, due to tissue-specific RNA processing, it is expressed as two different isoforms, localized to axon initial segments and nodes of Ranvier. Ankyrin-G is also found at sites of epithelial cell-cell contact and in the intracellular membranes of some cells. Ankyrin-G isoforms contain a unique stretch of sequence highly enriched in serine and threonine residues immediately following a globular head domain. Ankyrin-G is thought to participate in the maintece and targeting of ion channels and cell adhesion molecules to the nodes of Ranvier and to axon initial segments.Specifications
| ANK3 | |
| Polyclonal | |
| Unconjugated | |
| ANK3 | |
| ANK 3, ANK3, Ankyrin 3, ANKYRIN G | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 288 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q12955 | |
| ANK3 | |
| ANK3 Fusion Protein Ag26948 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction